Skip to Content
Merck
All Photos(1)

Key Documents

07-0050

Sigma-Aldrich

Decahydronaphthalene, mixture of cis + trans

SAJ special grade, ≥99.0%

Synonym(s):

Decalin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
878165
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ special grade

vapor density

4.76 (vs air)

vapor pressure

42 mmHg ( 92 °C)
741 mmHg ( 188 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

482 °F

expl. lim.

0.7-4.9 %, 100 °F

availability

available only in Japan

refractive index

n20/D 1.474 (lit.)

bp

189-191 °C (lit.)

mp

−125 °C (lit.)

density

0.896 g/mL at 25 °C (lit.)

SMILES string

C1CCC2CCCCC2C1

InChI

1S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10H,1-8H2

InChI key

NNBZCPXTIHJBJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Decalin is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Zhu et al.
Journal of natural products, 75(4), 567-571 (2012-03-08)
Five new decalin derivatives (1-5), together with two known compounds (6 and 7), were isolated from the ethyl acetate extract of red yeast rice. Their structures were elucidated by means of NMR and mass spectroscopic analyses. Monascusic lactone A (1)
Joseph B Lim et al.
The journal of physical chemistry. B, 116(1), 203-210 (2011-12-06)
A modification of the CHARMM36 lipid force field (C36) for cholesterol, henceforth, called C36c, is reported. A fused ring compound, decalin, was used to model the steroid section of cholesterol. For decalin, C36 inaccurately predicts the heat of vaporization (~10
Franco Cataldo et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 77(5), 998-1004 (2010-09-25)
Oleum (fuming sulphuric acid), a well known superacid, was used as medium for the generation of the radical cation of a series of selected PAHs. The resulting radical cation spectra were studied by electronic absorption spectroscopy. Not only common PAHs
Pranjal Chandra et al.
Biosensors & bioelectronics, 39(1), 139-144 (2012-08-14)
A highly sensitive and selective method is developed for the simultaneous detection of dopamine (DA) and acetaminophen (AP) by reactive blue-4 (RB4) dye entrapped poly1,5-diaminonaphthalne (polyDAN) composite film layer. The polyDAN-RB4 composite is electrochemically developed at glassy carbon electrode. The
Lei Chen et al.
Organic letters, 13(14), 3580-3583 (2011-06-16)
Regio-, stereo-, and facial selective [4 + 2] cycloadditions between highly activated vinyl sulfones and 1,3-dienes derived from (R)-4-tert-butyldimethylsilyloxy-2-cyclohexen-1-one provide a powerful approach for the asymmetric synthesis of compounds containing the bicyclo[2.2.2]octanone carbon skeleton. This new methodology has been successfully

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service