05-2753
Chlorine water
CP
Synonym(s):
Chlorine solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
CP
form
liquid
availability
available only in Japan
InChI
1S/Cl2/c1-2
InChI key
KZBUYRJDOAKODT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biochemistry, 52(14), 2482-2491 (2013-03-22)
A T203Y substitution in green fluorescent protein causes a red shift in emission to yield a class of mutants known as yellow fluorescent protein (YFP). Many of these YFP mutants bind halides with affinities in the millimolar range, which often
Journal of microbiology (Seoul, Korea), 51(1), 100-104 (2013-03-05)
The plant growth, stem sap flow, Na(+) and Cl(-) content, and the expression of vacuolar Na(+)/H(+) antiporter gene (LeNHX1) in the leaves and roots of tomato under different NaCl stresses (0.5% and 1%) were studied to analyze the effect of
American journal of physiology. Lung cellular and molecular physiology, 304(11), L765-L773 (2013-04-09)
Chlorine (Cl₂) is a highly irritating and reactive gas with potential occupational and environmental hazards. Acute exposure to Cl₂ induces severe epithelial damage, airway hyperreactivity, impaired alveolar fluid clearance, and pulmonary edema in the presence of heightened inflammation and significant
Carbohydrate polymers, 92(2), 2314-2320 (2013-02-13)
A biodegradable, β-cyclodextrin-based superabsorbent resin was synthesized by the inverse suspension method. The microstructure, chemical structure, and thermal performance of the resin were characterized by scanning electron microscopy, Fourier transform-infrared spectroscopy, and differential scanning calorimetry. The effects of the synthesis
Nanoscale, 5(5), 1945-1948 (2013-01-30)
We demonstrated the p-type chemical doping by chlorine and nitrate anions to enhance the Schottky junction in the solar cell. Nitrate ions were found to be more effective for reducing the sheet resistance and enlarging the work function of graphene
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service