Skip to Content
Merck
All Photos(2)

Key Documents

ZAFS0080WW

Milli-Q

AFS® Water Purification System

For analyzers with pure water needs up to 160 L/day

Synonym(s):

AFS® 8 Water Purification System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104206
NACRES:
SB.53

AC/DC input

100 - 230 V, 50 - 60 Hz

Quality Level

product line

AFS®

packaging

pkg of 1 unit

parameter

160 L/day max. usage
2 L/min distribution flow rate
28 kg operating weight (61.7 lb)
8 L/hr make-up flow rate

resistivity

>15 MΩ-cm, at 25 degrees

system H × W × depth

58.5 cm (23 in.) × 26.8 cm (10.6 in.) × 42.6 cm (16.8 in.)

total organic carbon (TOC) residue

<30 ppb

input

potable tap water

output

clinical laboratory reagent water (CLRW) (complies with the CLSI guideline)

General description

Analyzer Feed Systems (AFS®) provide a cost-effective solution for analyzers with pure water needs of up to 480 L daily. The AFS® 8 water purification system has a production flow rate to the reservoir of 8 L/h and a dispensing flow rate to the analyzer of up to 2 L/min. Water produced by AFS® systems complies with the Clinical and Laboratory Standards Institute (CLSI) guideline for clinical laboratory reagent water (CLRW). These systems combine complementary water purification techniques (complete pretreatment, advanced reverse osmosis (RO), bactericidal UV lamp, final 0.22 µm filtration) to provide reliable and consistent CLRW-quality water. AFS® systems have full monitoring and water quality archiving capabilities that allows storage of up to six months of data.

Application

AFS® water purification systems feed analyzers with consistent water quality that meets CLRW standards of the CLSI.

Features and Benefits

  • Complementary water purification techniques provide consistent water quality meeting CLRW standards
  • Cost-effective thanks to optimized lifetimes for pretreatment and polishing packs and low feed water use
  • Fulfills accreditation needs (e.g. CAP 15189SM accreditation to the ISO 15189:2022 standard) with full monitoring and automatic water quality archiving capabilities. Up to 6 months of information can be stored for reliable traceability
  • An intuitive icon-based system display facilitates maintenance, and an ergonomic pack-locking system makes pack changes easier than ever
  • Milli-Q® Service Plans offer a range of support, including preventive maintenance visits and online contract management
  • A small footprint allows installation wherever it’s convenient on the wall, on or under the bench, or on a cart

Components

  • The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
  • This catalog number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
  • Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!

Other Notes

Directions For Use
Organism Retention: Microorganisms
Mode of Action: Filtering and UV
Application: Clinical laboratories
Intended Use: Water purification
Instructions for Use: This item provides water filtered through 0.22 μm sterilizing-grade membranes and exposed to bactericidal UV lamp. Please refer to system equipment user guide section "System" (provided on USB key in the North American configuration version).
Storage Statement: Store in dry location
Disposal Statement: Dispose of in accordance with applicable federal, state, and local regulations.

Legal Information

AFS is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

Critical points for laboratories to consider when assessing bacteria levels in purified water and attempting to optimize the microbiology process.

See All

Related Content

Explore how high-quality water is essential for accurate clinical chemistry assays and complying with norms and guidelines (e.g. CLSI).

Explore how high-quality water is essential for accurate clinical chemistry assays and complying with norms and guidelines (e.g. CLSI).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service