Skip to Content
Merck
All Photos(1)

Key Documents

SCT032

Sigma-Aldrich

BioTracker 515 Green Zn2+ DA Dye

Live cell imaging dye for zinc (Zn2+) for cell signaling, protein self-assembly and cellular metabolism applications.

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.47

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

ambient

General description

Zinc (Zn2+) is a trace nutrient critical for physiological function. Although most biological zinc ions are tightly associated with proteins, pools of loosely bound or mobile forms serve regulatory or signaling functions, including nucleation of protein self-assembly, triggering of signaling pathways, and modification of cellular metabolism.

The BioTracker 515 Green Zn2+ Dye is a fluorescent imaging probe for the detection of Zn2+ in living cells. The BioTracker 515 Green Zn2+ Dye has high solubility, so that it is not permeable through the cell membrane. BioTracker 515 Green Zn2+ Dye DA has an additional acetyl group which make the reagent go inside the cell much easier, and the acetyl group is removed by the intracellular esterase which make the reagent stay inside the cell for extended periods of time.

Concentration of the provided sample is 5 mmol/L in DMSO. Dilute 1000 times with neutral buffer before use.

Spectral Properties
Absorbance: 492nm
Emission: 515nm

Application

Live cell fluorescent imaging
Research Category
Cell Imaging
Research Sub Category
Live Cell Dye

Physical form

Lyophilized

Storage and Stability

Store BioTracker 515 Green Zn2+ DA Dye at -20°C, desiccate and protect from light
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

849.6 °F

Flash Point(C)

454.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service