SCC449
OSUMMER.5 Mouse NRAS-Mutant Melanoma Cell Line
Mouse
Synonym(s):
Mouse melanoma cell line
Sign Into View Organizational & Contract Pricing
All Photos(6)
About This Item
UNSPSC Code:
41106514
NACRES:
NA.81
Recommended Products
product name
OSUMMER.5 Mouse NRAS-Mutant Melanoma Cell Line,
biological source
mouse
Quality Level
packaging
vial of ≥1x10˄6 cells
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
cell culture | mammalian: suitable
shipped in
liquid nitrogen
storage temp.
−196°C
Application
Each vial contains >=1X106 viable cells.
Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel and have been found to be negative for all pathologies in this test.
Cells are verified to be of mouse origin and negative for interspecies contamination from rat, human, Chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination Clear panel.
Cells are negative for mycoplasma contamination.
Cells are genotyped by STR analysis to verify the unique identity of the cell line.
Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel and have been found to be negative for all pathologies in this test.
Cells are verified to be of mouse origin and negative for interspecies contamination from rat, human, Chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination Clear panel.
Cells are negative for mycoplasma contamination.
Cells are genotyped by STR analysis to verify the unique identity of the cell line.
Melanoma is a skin cancer that develops in pigment-producing melanocytes. Sporadic genetic mutations have been associated with melanomas. Immunotherapy treatments targeting these genes or pathways have been proven to have significant benefits in melanoma patients by improving general response and survival.The OSUMMER (Ohio State University and Moffitt Melanoma Exposed to Radiation)cell lines fulfill a previously unfilled gap in melanoma biology. They are derived from the TN mouse model of melanoma, which are homozygous for the LSL-Nras Q61R allele. Oncogene expression is activated by tamoxifen-inducible Cre recombinase in melanocytes. Loss of function is an early event observed in the majority of human melanomas. TN mice additionally carry p16INK4A conditional knockout alleles. OSUMMER.5 NRAS mouse melanoma cell line was derived from a male TN mouse exposed to UVA and UVB radiation. OSUMMER.5 carries the specific NRAS mutation Q61R.WES revealed high SNV (Single Nucleotide variants) and low CNA (Copy Number Alterations) burden, as well as the presence of UV signature mutations. OSUMMER.5 cells express the melanoma markers PMEL and MelanA, and exhibit robust growth in culture.SourceOSUMMER.5 was derived from melanoma induced in TN (Tyr::CreER) mice exposed to UVA or UVB radiation on postnatal day 3.References1. Nat Rev Dis Primers 2015, 1: 150032.The Lancet, 392(10151), 971-984.3. Life Sci Alliance 2021, 4(9): e202101135.
Features and Benefits
OSUMMER.5 male mouse NRAS-mutant melanoma cell line carries the TN61R mutation and is derived from the TN mouse model. This model is syngeneic with the C57BL/6 mouse model. OSUMMER cells can form tumors in vivo without Matrigel injection.
Storage and Stability
OSUMMER.5 cells should be stored in liquid nitrogen until use. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.
Other Notes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service