Skip to Content
Merck
All Photos(1)

Key Documents

RU5070

Sigma-Aldrich

Rabies Antibody Diluent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

antibody form

purified antibody

Quality Level

antibody product type

primary antibodies

clone

monoclonal

packaging

antibody small pack of 100 mL

shipped in

2-8°C

General description

Rabies Diluent Reagent is intended for use as a dilution buffer for titrating Anti-Rabies Blend, clones 103.7/502.2, FITC (Cat. No. RU5500).

Application

Meets specifications.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Physical form

Purified in PBS containing 1% BSA, 0.05% Tween-20 and 0.1% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Please refer to the Certificate of

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service