Skip to Content
Merck
All Photos(1)

Key Documents

PX1570

Supelco

Potassium phosphate dibasic

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Potassium Hydrogen Phosphate, Dipotassium Phosphate, Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-834-5

Agency

meets USP/NF

Quality Level

Assay

≥98.0% (acidimetric)

form

granular

impurities

≤0.01% Insoluble matter

loss

≤1.0% loss on drying, 105°C

color

white

pH

9 (20 °C, 10 g/L in H2O)

solubility

1600 g/L

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
N: ≤0.001%
Na: ≤0.05%
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

ZPWVASYFFYYZEW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (acidimetric)98.0% minChloride (Cl)0.003% maxColorWhiteFormGranular powderHeavy Metals (by ICP-OES)5ppm maxInsoluble matter0.01% maxIron (Fe)0.001% maxLoss on drying (105°C)1.0% maxNitrogen compounds (as N)0.001% maxSodium (Na)0.05% maxSulfate (SO4)0.005% maxpH of a 5% solution at 25°C8.5-9.6

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service