Skip to Content
Merck
All Photos(1)

Key Documents

PX0046

Supelco

Paraffin Oil

heavy

Synonym(s):

Paraffin Oil, Mineral Oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502
EC Index Number:
232-384-2
Pricing and availability is not currently available.

form

liquid

color

APHA: ≤20

shipped in

ambient

storage temp.

room temp

General description

Heavy

Application

Heavy
Saybolt viscosity 310 at 100F

Analysis Note

Color (APHA)20 maxDescriptionTo pass testFormLiquidInfrared SpectrumConforms to standardLimit of polynuclear compoundsTo pass testNeutralityTo pass testReadily carbonizable substancesTo pass testSolid paraffinTo pass testSolubilityTo pass testSpecific Gravity at 25/25°C0.855-0.880

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service