Skip to Content
Merck
All Photos(1)

Documents

PRCYTA-40K-BK38

Millipore

MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A 38-Plex Premixed Magnetic Bead Panel - BULK (Space Saver) PACKAGING

Synonym(s):

Cytokine panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.47

species reactivity

nonhuman primates

assay range

accuracy: 80-102%
inter-assay cv: <20%
intra-assay cv: <10%

inter-assay cv: <30%
(TNF-α)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

“Cytokine” is a general term used for a diverse group of small proteins, peptides or glycoproteins secreted by lymphocytes, monocytes, macrophages and other cells that regulate immune responses, haematopoiesis and lymphocyte development. Cytokines include interleukins, interferons, chemokines and other signaling molecules. Growth factors are extracellular polypeptides that have a positive effect on cell growth and proliferation. They can stimulate growth in a variety of different cell types. Growth factors and cytokines are similar in structure and the way of action. Individual cytokines or growth factors are produced by multiple cell types. This is different from hormones, which tend to be made by specialized glands. Each cytokine or growth factor acts through its own receptor on target cells to generate signaling pathways, and as a consequence to regulate biological processes. Some intracellular signaling components are shared between cytokines and growth factors. Expression of cytokines or growth factors and their receptors is highly regulated. De-regulation may contribute to many diseases such as infectious disease, autoimmune and chronic inflammatory disease, cardiovascular disease, metabolic syndrome, neurological disorders, and cancer. Cytokine and growth factor research plays a significant role in achieving a deeper understanding of the immune system and combating related diseases. As the most closely related species to humans, non-human primates are critical models for those studies.
The MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A 38-Plex Premixed Magnetic Bead Panel is a 38-plex kit to be used for the simultaneous quantification of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: sCD137, CD40L, sFASL, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, VEGF-A.

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel, except G-CSF had ∼5% cross-reactivity with IL-1β.

Application

  • Analytes: sCD137, CD40L, sFASL, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, VEGF-A
  • Recommended Sample type: Serum, plasma or tissue/cell lysate and culture supernatant samples
  • Recommended Sample dilution: Neat plasma or serum. Tissue culture supernatant may require a dilution with an appropriate control medium prior to assay. RANTES requires a 1:100 dilution of plasma/serum samples.

NOTE: RANTES cannot be plexed with other cytokines in this panel due to a required 1:100 dilution of plasma/serum samples. This premixed panel comes with a RANTES bead, so please plan your samples accordingly.
Note: The serum/plasma samples of some non-human primate species may require up to 1:100 dilution in Assay Buffer for IL-8.
  • Assay Run Time: Overnight or two-hour primary incubation. For best results, an overnight incubation is recommended
  • Research Category: Inflammation & Immunology
  • Research Sub Category: Obesity, Metabolic Disorders, Inflammation & Autoimmune Mechanisms

Features and Benefits

Configuration: Premixed

Storage and Stability

  • Recommended storage for kit components is 2 - 8°C.
  • For long-term storage, freeze reconstituted standards and controls at -20°C. Avoid multiple freeze/thaw cycles.
  • DO NOT FREEZE Antibody-Immobilized Beads, Detection Antibody, and Streptavidin-Phycoerythrin

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MILLIPLEX® Non-Human Primate Panel A allows comprehensive immune factor profiling on a single 96-well plate.

MILLIPLEX® Non-Human Primate Panel A allows comprehensive immune factor profiling on a single 96-well plate.

MILLIPLEX® Non-Human Primate Panel A allows comprehensive immune factor profiling on a single 96-well plate.

MILLIPLEX® Non-Human Primate Panel A allows comprehensive immune factor profiling on a single 96-well plate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service