PLGC07610
Regenerated Cellulose Membrane Filter, 10 kDa NMWCO
Ultracel®, filter diam. 76 mm
Synonym(s):
Ultrafiltration Discs, 10 kDa NMW, Ultrafiltration membrane discs
About This Item
Recommended Products
material
regenerated cellulose membrane
Quality Level
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
Ultracel®
technique(s)
protein clean-up: suitable (desalting)
protein purification: suitable (concentration)
diam.
76 mm
filter diam.
76 mm
filtration area
41.8 cm2
matrix
Ultracel®
pore size
10 kDa NMWCO
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore PVDF cut discs are available 0.1 µm, 0.22 µm, and 0.45 µm pore sizes in both 63.5 mm and 75 mm diameters.
Features & Benefits:
•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
•Both Ultracel, Durapore, and Biomax membranes are available in a wide range of pore sizes to meet your separation requirements
•Compatible with Amicon Stirred Cells
Applications:
Concentration, Buffer Exchange, Desalting
Application
Linkage
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service