OX0101
1-Octanesulfonic Acid, Sodium Salt, Monohydrate
suitable for HPLC
Synonym(s):
1-Octanesulfonic Acid, Sodium Salt, Monohydrate, Octanesulfonic Acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
solid
Quality Level
technique(s)
HPLC: suitable
shipped in
ambient
storage temp.
room temp
InChI
1S/C8H18O3S.Na.H2O/c1-2-3-4-5-6-7-8-12(9,10)11;;/h2-8H2,1H3,(H,9,10,11);;1H2/q;+1;/p-1
InChI key
MBURIAHQXJQKRE-UHFFFAOYSA-M
General description
1-Octanesulfonic Acid, Sodium Salt, Monohydrate
OX0101-3 100g Poly Bottle
OX0101-3 100g Poly Bottle
HPLC
Application
HPLC
Ion Pair Reagent
Analysis Note
Color: White
Form: Solid
Purity: 99% min
Solubility in water (5% w/v): To pass test
UV Abs. at 210 nm: 1.00AU max
UV Abs. at 220 nm: 0.10AU max
UV Abs. at 254 nm: 0.06AU max
UV Abs. at 280 nm: 0.06AU max
Form: Solid
Purity: 99% min
Solubility in water (5% w/v): To pass test
UV Abs. at 210 nm: 1.00AU max
UV Abs. at 220 nm: 0.10AU max
UV Abs. at 254 nm: 0.06AU max
UV Abs. at 280 nm: 0.06AU max
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service