Key Documents
MZPNNVX00
Millipore
Multiscreen® 384 well Plate
V-bottom clear natural polypropylene plate, non-sterile, pkg of 100 plates
About This Item
UNSPSC Code:
41122102
NACRES:
NB.15
material:
V-bottom clear natural polypropylene plate
plate size:
384 wells
sterility:
non-sterile
material
V-bottom clear natural polypropylene plate
Quality Level
sterility
non-sterile
product line
MultiScreen®
packaging
pkg of 100 plates
plate size
384 wells
General description
MultiScreen® 384-well polypropylene microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices. Polypropylene microplates are appropriate for sample preparation and storage in addition to some biochemical assays.
Physical form
Color Code:Natural
Other Notes
Automation Compatible:Yes
Legal Information
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany