Skip to Content
Merck
All Photos(3)

Key Documents

MRCF0R100ET

Millipore

Microcon® DNA Fast Flow Centrifugal Filters

sample volume 0.5 mL, Ultracel® regenerated cellulose membrane (low binding), Ethylene Oxide (EtO) treated

Synonym(s):

Centrifuge concentrator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24
Pricing and availability is not currently available.

material

Ultracel® regenerated cellulose membrane (low binding)
polyacetal base (membrane support)
polycarbonate device top
polypropylene tube (for filtrate/concentrate)
silicone O-ring (medical-grade, dual cycle ethylene oxide (EtO) treated)

Quality Level

sterility

non-sterile

feature

holdup volume> 10 μL

packaging

pkg of 20 ea

manufacturer/tradename

Microcon®

parameter

0.5 mL sample volume

technique(s)

DNA purification: suitable
RNA purification: suitable

L

4.50 cm (1.8 in.)

diam.

12.3 mm

filtration area

0.32 cm2

working volume

0.5 mL

shipped in

ambient

General description

Filter Type: Ultrafiltration
Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes. With the low-binding Ultracel membrane, Microcon filters offer:
•Typical recoveries of greater than 95%, even for dilute solutions
•Reverse spin to maximize recovery, even in the smallest samples
•Convenient storage of filtrate or concentrated sample in standard microfuge tube
•Concentration factors up to 100X

Microcon DNA Fast Flow Filter
Optimized for the concentration and recovery of genomic DNA with SDS buffer. The low nonspecific binding characteristics of the membrane and the other device components, coupled with its medical-grade o-ring seal, allows the device to accommodate several wash steps with minimal sample loss.

Features:
•High recovery for small volumes with reverse spin (concentration factor less than 20X)
•Low-binding Ultracel membrane
•Fast processing

Sample applications:
•Recovery of genomic DNA for forensic applications
•Concentration and desalting of nucleic acids (high recovery alternative to ethanol precipitation)
•Removal of primers from amplified DNA
•Easy filtrate recovery, exploiting the flat membrane design

Features and Benefits

Minimum Final Concentrate Volume: 50 µL

Physical form

Color Code: Blue

Legal Information

MICROCON is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Microcon® Centrifugal Filters simply and efficiently concentrate and desalt solutions of DNA, RNA, protein or other macromolecules, using any centrifuge that can accept 1.5 mL tubes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service