Skip to Content
Merck
All Photos(4)

Key Documents

MC96U6

Millipore

Millicell® Microwell Plates

600 µm U-bottom 96-well Hydrogel Plates, 5 pack, lid, skirt, Organoid and Spheroid Culture

Synonym(s):

3D Culture plates, microplates, microwell plates, multiwell plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24

material

polystyrene
round clear plastic bottom

Quality Level

description

Organoid and Spheroid Culture
PEG Hydrogel

feature

lid
skirt
Number of Microwells- 55 (may vary up to 7% fewer)
specific plates for 3D cell culture

manufacturer/tradename

Millicell®

packaging

pack of 5 ×

plate size

96 well

size

600 μm , microwell

working volume

200 μL

General description

Millicell Microwell Plates are a read-to-use platform for high-throughput and reproduciblity of organoid cultures. Based on an array of ultra-dense U-bottom shaped microwells (uW) in hydrogel, single organoids are robustly generated in each microcvity and grown in suspension-like culture without a solid extracellular matrix. With mulitple configurations available, there is a microwell size that will fit your organoid model.

Application

Organoid and Spheroid Culture

Features and Benefits

  • O-ring with meniscus breaking features: For homogeneous seeding
  • Independent port: For medium change without disturbing the organoids
  • Multiple microwell sizes: Accommodate any organoid type
  • Biocompatible and cell repellent: For efficient aggregation of stem cells
  • Water-based: Allowing diffraction-less imaging
  • Tunable chemistry: For customization according to organoid needs
  • No solid matrix needed

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Learn how to cultivate similar-sized iPSC-derived colon organoids using Millicell® Microwell plates and perform a forskolin-induced swelling assay.

Learn how to cultivate similar-sized iPSC-derived colon organoids using Millicell® Microwell plates and perform a forskolin-induced swelling assay.

Related Content

Explore our preclinical models, tools for 2D and 3D cell culture, cell-based assays, reagents, instruments, and more to support antibody drug development.

Explore our preclinical models, tools for 2D and 3D cell culture, cell-based assays, reagents, instruments, and more to support antibody drug development.

Explore our preclinical models, tools for 2D and 3D cell culture, cell-based assays, reagents, instruments, and more to support antibody drug development.

Tools to accelerate preclinical studies in antibody drug development, from toxicity to immunogenicity, pharmacokinetics, pharmacodynamics, and efficacy.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service