Skip to Content
Merck
All Photos(2)

Key Documents

MABS1889

Sigma-Aldrich

Anti-FANCD2 Antibody, clone 10F8.1

clone 10F8.1, from mouse

Synonym(s):

FFanconi anemia group D2 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
10F8.1, monoclonal
application:
IHC
WB
species reactivity:
human
technique(s):
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10F8.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... FANCD2(2177)

General description

Fanconi anemia group D2 protein (UniProt: Q9BXW9; also known as Protein FACD2) is encoded by the FANCD2 (also known as FACD) gene (gene ID: 2177) in human. FACD2 is a nuclear protein that is required for maintenance of chromosomal stability. It is highly expressed in fetal oocytes, and in hematopoietic cells of the fetal liver and bone marrow. High expression is also observed in germinal center cells of the spleen, tonsil, and reactive lymph nodes and in the proliferating basal layer of squamous epithelium of tonsil, esophagus, larynx and cervix. FACD2 promotes accurate and efficient pairing of homologs during meiosis. It is also involved in the repair of DNA double-strand breaks, both by homologous recombination and single-strand annealing and may participate in S phase and G2 phase checkpoint activation upon DNA damage. It is concentrated in nuclear foci during S phase and upon genotoxic stress. FACD2 plays a role in preventing breakage and loss of missegregating chromatin at the end of cell division, particularly after replication stress. It is phosphorylated in response to various genotoxic stresses by ATM and/or ATR. Upon ionizing radiation, it is phosphorylated on serine 222 and serine 1404 by ATM. Phosphorylation on serine 222 is shown to be essential for S-phase checkpoint activation. Mutations in FANCD2 gene are known to cause Fanconi anemia complementation group D2, which results in anemia, leukopenia, and thrombopenia and may cause cardiac, renal, and limb malformations and increase predisposition to malignancies. (Ref.: Shen, Y., et al. (2015). Cancer Cell Microenviron. 2(4): e986).

Specificity

Clone 10F8.1 specifically detects human Fanconi anemia group D2 protein. It targets an epitope with in the first 150 amino acids from the N-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to first 150 amino acids of human Fanconi anemia group D2 protein.

Application

Anti-FANCD2, clone 10F8.1, Cat. No. MABS1889, is a mouse monoclonal antibody that detects Fanconi anemia group D2 protein and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected FANCD2 in human tonsil and human testis tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected FANCD2 in Jurkat cell lysate.

Target description

~166 kDa observed; 164.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service