Skip to Content
Merck
All Photos(2)

Key Documents

MABN77

Sigma-Aldrich

Anti-Potassium Channel Kv1.2 Antibody, clone K14/16

clone K14/16, from mouse

Synonym(s):

potassium voltage-gated channel, shaker-related subfamily, member 2, potassium voltage-gated channel subfamily A member 2, Voltage-gated potassium channel subunit Kv1.2, Voltage-gated potassium channel HBK5, potassium channel, Voltage-gated K(+) channel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

K14/16, monoclonal

species reactivity

rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNA2(3737)

General description

Potassium channel Kv1.2 is one of many different types of potassium channels in the cell. The Kv family of potassium channels seem to have a conserved homotetramer formation consisting of four voltage sensors and one pore domain. Kv1.2 cooperates with cortactin (an actin cytoskeleton-binding protein) and together they may have a part in the regulation of the ionic current.

Immunogen

Recombinant protein corresponding to rat Kv1.2.

Application

Immunofluorescence Analysis: a previous lot of this antibody was used by an independent laboratory in IF. (Yang, J.W., et al. (2007). PNAS. 104(50):20055–20060.)
This Anti-Potassium Channel Kv1.2 Antibody, clone K14/16 is validated for use in IH, WB, IF for the detection of Potassium Channel Kv1.2.

Quality

Evaluated by Western Blot in rat brain membrane tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Kv1.2 on 10 µg of rat brain membrane tissue lysate.

Target description

~ 65 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María T Dours-Zimmermann et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(24), 7731-7742 (2009-06-19)
The CNS-restricted versican splice-variant V2 is a large chondroitin sulfate proteoglycan incorporated in the extracellular matrix surrounding myelinated fibers and particularly accumulating at nodes of Ranvier. In vitro, it is a potent inhibitor of axonal growth and therefore considered to
De-En Xu et al.
Cell adhesion & migration, 8(4), 396-403 (2014-12-09)
Amyloid precursor protein (APP), commonly associated with Alzheimer disease, is upregulated and distributes evenly along the injured axons, and therefore, also known as a marker of demyelinating axonal injury and axonal degeneration. However, the physiological distribution and function of APP
Yoshinori Otani et al.
Communications biology, 3(1), 121-121 (2020-03-15)
Charcot-Marie-Tooth (CMT) disease is a hereditary neuropathy mainly caused by gene mutation of peripheral myelin proteins including myelin protein zero (P0, MPZ). Large myelin protein zero (L-MPZ) is an isoform of P0 that contains an extended polypeptide synthesized by translational
O W Howell et al.
Brain : a journal of neurology, 129(Pt 12), 3173-3185 (2006-10-17)
Saltatory conduction in the nervous system is enabled through the intimate association between the leading edge of the myelin sheath and the axonal membrane to demarcate the node of Ranvier. The 186 kDa neuron specific isoform of the adhesion molecule
Du-Yu Nie et al.
The EMBO journal, 22(21), 5666-5678 (2003-11-01)
We report Nogo-A as an oligodendroglial component congregating and interacting with the Caspr-F3 complex at paranodes. However, its receptor Nogo-66 receptor (NgR) does not segregate to specific axonal domains. CHO cells cotransfected with Caspr and F3, but not with F3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service