Skip to Content
Merck
All Photos(2)

Key Documents

MABC83

Sigma-Aldrich

Anti-Vps4/SKD1 Antibody, clone 5E6.1

clone 5E6.1, from mouse

Synonym(s):

Vacuolar protein sorting-associated protein 4B, Cell migration-inducing gene 1 protein, Suppressor of K(+) transport growth defect 1, Protein SKD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5E6.1, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... VPS4B(9525)

General description

Vacuolar protein sorting-associated protein 4B (Vps4), or SKD1, is a ubiquitous AAA ATPase involved in endosomal vesicle trafficking. Trafficking of vesicles containing proteins such as receptors, from the endosome to the cytoplasm, is an important regulatory mechanism for cell-surface receptors and the signaling pathways that involve those receptors. Such endosomal vesicles are shuttled from the endosome to the cytoplasm by multivesicular bodies (MVB). MVBs are large multi-protein complexes that assemble and disassemble during this process. Previous studies have indicated that the yeast homologue of Vps4, Vps4p (60 % sequence identity to Vps4), interacts with endosomes via its N-terminal microtubule interacting and trafficking (MIT) domain, and also associates with a number of MVB proteins including Vps2p, Bro1p, Vps20p, Vta1p, and Did2p/Chm1p. Although the precise role of Vps4 is still unclear, previous studies have suggested that it interacts with the ESCRT-III complex involved in the sorting of MVBs and has been associated with virus-loaded vesicles such those in HIV-infected cells.

Immunogen

Linear peptide corresponding to human Vps4/SKD1.

Application

Anti-Vps4/SKD1 Antibody, clone 5E6.1 is an antibody against Vsp4/SKD1 for use in Western Blotting, ICC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Vps4/SKD1 in NIH/3T3, A431, and HeLa cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blot in human skeletal muscle tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Vps4/SKD1 in 10 µg of human skeletal muscle tissue lysate.

Target description

~52 kDa observed. The calculated molecular weight of this protein is ~50 kDa Uniprot describes that this protein is autophosphoralated upon DNA damage.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human skeletal muscle tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service