Skip to Content
Merck
All Photos(1)

Key Documents

HX0636

Supelco

Hydrogen Peroxide

OmniTrace Ultra

Synonym(s):

Hydrogen peroxide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
41116105
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

vapor density

1.2 (vs air)

vapor pressure

18 hPa ( 20 °C)
23.3 mmHg ( 30 °C)

form

liquid

concentration

≥30-35% in hydrogen peroxide

pH

2-4 (20 °C in H2O)

bp

108 °C/1013 hPa

mp

-25.5 °C

solubility

soluble (Water: Soluble)

density

1.11 g/cm3 at 20 °C (68 °F)

shipped in

ambient

storage temp.

no temp limit

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • Stable Pentagonal Layered Palladium Diselenide Enables Rapid Electrosynthesis of Hydrogen Peroxide.: This paper presents a novel material, palladium diselenide, which enables rapid electrosynthesis of hydrogen peroxide. The findings have implications for sustainable hydrogen peroxide production and electrochemical applications (Ross et al., 2024).


  • Green-synthesized zinc oxide nanoparticles by Enterobacter sp.: unveiling characterization, antimicrobial potency, and alleviation of copper stress in Vicia faba (L.) plants.: This study explores the green synthesis of zinc oxide nanoparticles using Enterobacter sp., emphasizing their antimicrobial properties and ability to alleviate copper stress in plants. Hydrogen peroxide is discussed as a critical agent in the nanoparticle synthesis and stress mitigation process (Elsilk et al., 2024).


  • Use of Raman spectroscopy and PCA for quality evaluation and out-of-specification identification in biopharmaceutical products.: This research utilizes Raman spectroscopy and Principal Component Analysis (PCA) to evaluate the quality of biopharmaceutical products. The study discusses the detection of hydrogen peroxide as a degradation product, crucial for ensuring product quality and compliance (Massei et al., 2024).



Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

ICP-MS analyzes 12 heavy metals in hemp-containing food samples using standard addition calibration for accurate quantification.

Questions

  1. HX0636 IS THIS 100% HYDROGEN PEROXIDE

    1 answer
    1. The concentration of this product is 30-32 % in water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service