EZFITBASE1
EZ-Fit® Manifold
316L stainless steel, 1-place manifold base for filtration application
Synonym(s):
EZ-Fit® Manifold base, EZ-Fit® manifold base for EZ-Stream® vacuum pump
About This Item
Recommended Products
Quality Level
material
316L stainless steel
description
1-place manifold base for filtration application, stainless steel
Sterilization Compatibility
autoclavable compatible
feature
1-place manifold base for filtration application
packaging
pkg of 1 unit
manufacturer/tradename
EZ-Fit®
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
for use with all EZ-Fit® Filtration heads (EZ-Fit®, Microfil®, Milliflex Oasis®)
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Legal Information
accessory
configured for
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service