The molar extinction coefficient of the product at 410 nm is estimated to be approximately 35,000 M-1 cm-1. This value is slightly lower than the 36,000 M-1 cm-1 (415 - 420 nm) reported in the publications below:
Appl. Environ. Microbiol. 2014, 80(24), 7484-7495
W.G. Biochem. J. 1975, 145, 93
ES004
ABTS Single Reagent, Blue green color,Horseradish Peroxidase Substrate (soluble)
500 ml ABTS Single Reagent, Blue green color, Horseradish Peroxidase Substrate (soluble) for ELISA.
Synonym(s):
Horseradish Peroxidase Substrate
About This Item
Recommended Products
form
liquid
manufacturer/tradename
Chemicon®
technique(s)
ELISA: suitable
λmax
410 nm
650 nm
General description
Application
Physical form
Storage and Stability
Legal Information
Disclaimer
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Mit welchem Extinktionskoeffizienten kann für dieses Produkt bei 410 nm gerechnet werden?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service