Skip to Content
Merck
All Photos(1)

Key Documents

DX1729

Supelco

N,N-Dimethylformamide

Synonym(s):

Formic Acid Dimethylamide, FM approval, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-679-5
Assay:
≥99.9% (GC)
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

833 °F

shelf life

24 mo. (from manufacture date)

expl. lim.

15.2 %

impurities

≤0.5 μeq/g Titrable acid
≤0.5 μeq/g Titrable base
≤300 ppm Water (H2O)
≤5 ppm Free amine (as dimethylamine)

evapn. residue

≤5 ppm

color

clearAPHA: ≤10

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

density

0.944 g/mL (lit.)

UV cutoff

≤270 nm

UV absorption

λ: 270 nm Amax: ≤1.00
λ: 275 nm Amax: ≤0.30
λ: 280 nm Amax: ≤0.20
λ: 300 nm Amax: ≤0.05
λ: 350 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solvent for many hydrophobic organic compounds.

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Expiration dateExp 24 months from mfg dateAppearanceClear, free from particulatesAssay (GC)99.9% minColor (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormLiquidFree amine (as dimethylamine)5ppm maxTitrable acid0.5µeq/g maxTitrable base0.5µeq/g maxUV Abs. at 270 nm1.00AU maxUV Abs. at 275 nm0.30AU maxUV Abs. at 280 nm0.20AU maxUV Abs. at 300 nm0.05AU maxUV Abs. at 350 nm0.01AU maxUV Cut-off270nm maxWater (H2O)300ppm max

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service