CMTI-2
EmbryoMax® Embryonic Stem Cell Line - Strain C57/BL6, passage 11, normal male genotype
Synonym(s):
BRUCE-4 ES cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76
Recommended Products
product line
EmbryoMax®
technique(s)
cell culture | stem cell: suitable
General description
Embryonic Stem Cell Lines derived from murine strain C57/BL6J selected for high targeting efficiency and proven germ line transmission. Cells are supplied as 2.5 x 106 ES cells per vial (approximately 2.5 population doublings per passage, shipped frozen on dry ice) and are supplied at passage 11.
Application
These Embryonic Stem Cell Lines are derived from murine strain C57/BL6J & selected for high targeting efficiency & proven germ line transmission.
Other Notes
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Legal Information
EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service