Skip to Content
Merck
All Photos(1)

Key Documents

AB5716

Sigma-Aldrich

Anti-Hey 2/HRT2 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... HEY2(23493)

Specificity

Recognizes Hey 2 / HRT2, a Helix-loop-helix class of transcription factor.

Immunogen

Synthetic peptide, amino acids 314-330 of human Hey 2 / HRT2.

Application

Anti-Hey 2/HRT2 Antibody is an antibody against Hey 2/HRT2 for use in IH.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of HES and HEY genes in infantile hemangiomas.
Adepoju, O; Wong, A; Kitajewski, A; Tong, K; Boscolo, E; Bischoff, J; Kitajewski, J; Wu, JK
Vascular Cell null
Sameer H Halani et al.
NPJ precision oncology, 2, 24-24 (2018-11-13)
Oligodendrogliomas are diffusely infiltrative gliomas defined by IDH-mutation and co-deletion of 1p/19q. They have highly variable clinical courses, with survivals ranging from 6 months to over 20 years, but little is known regarding the pathways involved with their progression or
Madhumita Basu et al.
JCI insight, 2(20) (2017-10-20)
Birth defects are the leading cause of infant mortality, and they are caused by a combination of genetic and environmental factors. Environmental risk factors may contribute to birth defects in genetically susceptible infants by altering critical molecular pathways during embryogenesis
Ana-Rita Pedrosa et al.
The Prostate, 76(1), 80-96 (2015-10-01)
The Notch signaling pathway has been implicated in prostate development, maintenance and tumorigenesis by its key role in cell-fate determination, differentiation and proliferation. Therefore, we proposed to analyze Notch family members transcription and expression, including ligands (Dll1, 3, 4 and
Danielle Daley-Brown et al.
Disease markers, 2017, 8248175-8248175 (2017-07-01)
The expression of NILCO molecules (Notch, IL-1, and leptin crosstalk outcome) and the association with obesity were investigated in types I and II endometrial cancer (EmCa). Additionally, the involvement of NILCO in leptin-induced invasiveness of EmCa cells was investigated. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service