Skip to Content
Merck
All Photos(1)

Key Documents

8.21944

Sigma-Aldrich

Alkylbenzyldimethylammonium chloride

for synthesis

Synonym(s):

Alkylbenzyldimethylammonium chloride, Benzyldimethylalkylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[RN(CH3)2CH2C6H5]Cl
UNSPSC Code:
12352107
NACRES:
NA.22

Quality Level

form

paste

pH

6-8 (20 °C, 10 g/L in H2O)

mp

37-41 °C

bulk density

800 kg/m3

storage temp.

2-30°C

Analysis Note

Assay (argentometric, calc. on anhydrous substance): 97.0 - 103.0 %
Water (K. F.): 4.0 - 6.0 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service