Skip to Content
Merck
All Photos(1)

Key Documents

8.20242

Sigma-Aldrich

Dibutyl disulfide

for synthesis

Synonym(s):

Dibutyl disulfide, Butyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3SS(CH2)3CH3
CAS Number:
Molecular Weight:
178.36
MDL number:
UNSPSC Code:
12352113
EC Index Number:
211-091-3
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥97.0% (GC)

form

liquid

expl. lim.

0.8 % (v/v)

mp

-94 °C

transition temp

flash point 93 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

S(SCCCC)CCCC

InChI

1S/C8H18S2/c1-3-5-7-9-10-8-6-4-2/h3-8H2,1-2H3

InChI key

CUDSBWGCGSUXDB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.936 - 0.938
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service