Skip to Content
Merck
All Photos(1)

Documents

8.18894

Sigma-Aldrich

Magnesium ethylate

for synthesis

Synonym(s):

Magnesium ethylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10MgO2
CAS Number:
Molecular Weight:
114.43
MDL number:
UNSPSC Code:
12352107
EC Index Number:
219-323-5
NACRES:
NA.22

Quality Level

Assay

≥97.0% (acidimetric)

form

granular

autoignition temp.

70 °C

potency

>3160 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Grignard Reaction

bulk density

480 kg/m3

storage temp.

2-30°C

InChI

1S/2C2H5O.Mg/c2*1-2-3;/h2*2H2,1H3;/q2*-1;+2

InChI key

XDKQUSKHRIUJEO-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 97.0 %
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-heat. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service