8.18629
n-Undecane
for synthesis
Synonym(s):
n-Undecane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)9CH3
CAS Number:
Molecular Weight:
156.31
MDL number:
UNSPSC Code:
12352001
EC Index Number:
214-300-6
NACRES:
NA.22
Pricing and availability is not currently available.
vapor pressure
0.6 hPa ( 20 °C)
Quality Level
Assay
≥99.0% (GC)
form
liquid
autoignition temp.
220 °C
expl. lim.
0.6-6.5 % (v/v)
kinematic viscosity
1.6 cSt(20 °C)
mp
-26 °C
transition temp
flash point 60 °C
density
0.74 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
C(CCCCC)CCCCC
InChI
1S/C11H24/c1-3-5-7-9-11-10-8-6-4-2/h3-11H2,1-2H3
InChI key
RSJKGSCJYJTIGS-UHFFFAOYSA-N
Application
- Reactive Extraction: The study explores the reactive extraction efficiencies of lactic acid and other organic acids using n-undecane in a solvent system (N Mungma, M Kienberger, M Siebenhofer - ChemEngineering, 2019).
- Plasma Catalytic Degradation: Research on using cerium oxide-supported catalysts for the plasma catalytic degradation of n-undecane to reduce emissions from the printing industry (T Xia, S Yao, Z Wu, G Li, J Li - Journal of Hazardous Materials, 2022).
Analysis Note
Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.739 - 0.741
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 0.739 - 0.741
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 3
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
140.0 °F
Flash Point(C)
60 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service