8.18557
(Chloromethyl)trimethylsilane
for synthesis
Synonym(s):
(Chloromethyl)trimethylsilane, Trimethylsilylchloromethane
About This Item
Quality Level
Assay
≥98.0% (GC)
transition temp
flash point 3 °C
density
0.88 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
[Si](CCl)(C)(C)C
InChI
1S/C4H11ClSi/c1-6(2,3)4-5/h4H2,1-3H3
InChI key
OOCUOKHIVGWCTJ-UHFFFAOYSA-N
Related Categories
Analysis Note
Density (d 20 °C/ 4 °C): 0.882 - 0.884
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
24.8 °F - closed cup
Flash Point(C)
-4 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service