8.18212
1,3-Dimethyl-2-imidazolidinone
for synthesis
Synonym(s):
1,3-Dimethyl-2-imidazolidinone, Dimethylethylene urea, DMI, DMEU
About This Item
vapor pressure
<1 hPa ( 20 °C)
Quality Level
Assay
≥99.0% (GC)
form
liquid
autoignition temp.
305 °C
potency
1261 mg/kg LD50, oral (Rat)
986 mg/kg LD50, skin (Rabbit)
expl. lim.
1.3-8.4 % (v/v)
kinematic viscosity
1.43 cSt(40 °C)
mp
8 °C
transition temp
flash point 114 °C
density
1.056 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
N1(CCN(C1=O)C)C
InChI
1S/C5H10N2O/c1-6-3-4-7(2)5(6)8/h3-4H2,1-2H3
InChI key
CYSGHNMQYZDMIA-UHFFFAOYSA-N
Application
- Concentration of 1, 3-dimethyl-2-imidazolidinone in aqueous solutions by sweeping gas membrane distillation: from bench to industrial scale. This study explores the characteristics and applications of 1,3-dimethyl-2-imidazolidinone (DMI) as a dipolar aprotic solvent, between tetrahydrofuran (THF) and hexamethylphosphoramide (HMPA) (R Abejón, et al., 2019).
- Photocatalyzed iron-based ATRP of methyl methacrylate using 1, 3-dimethyl-2-imidazolidinone as both solvent and ligand. This paper discusses the use of DMI in green solvent applications, highlighting its dual role as a solvent and ligand in polymerization processes (J Wu, et al., 2017).
- Perovskite solar cells fabricated by using an environmental friendly aprotic polar additive of 1, 3-dimethyl-2-imidazolidinone. The study showcases the application of DMI in improving the quality of perovskite films for solar cells (L Zhi, et al., 2017).
Analysis Note
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 1.055 - 1.057
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
219.2 °F - closed cup
Flash Point(C)
104 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service