Skip to Content
Merck
All Photos(1)

Key Documents

8.00784

Sigma-Aldrich

1,2,4,5-Benzenetetracarboxylic acid

for synthesis

Synonym(s):

1,2,4,5-Benzenetetracarboxylic acid, Benzene-1,2,4,5-tetracarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H2(COOH)4
CAS Number:
Molecular Weight:
254.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-879-5
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

mp

250-260 °C

solubility

15 g/L

density

1.79 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)c1c(cc(c(c1)C(=O)O)C(=O)O)C(=O)O

InChI

1S/C10H6O8/c11-7(12)3-1-4(8(13)14)6(10(17)18)2-5(3)9(15)16/h1-2H,(H,11,12)(H,13,14)(H,15,16)(H,17,18)

InChI key

CYIDZMCFTVVTJO-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

617.0 °F

Flash Point(C)

325 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service