Skip to Content
Merck
All Photos(1)

Key Documents

8.00503

Sigma-Aldrich

Butyric anhydride

for synthesis

Synonym(s):

Butyric anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CO)2O
CAS Number:
Molecular Weight:
158.19
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-383-4
NACRES:
NA.22

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

290 °C

potency

8790 mg/kg LD50, oral (Rat)
6400 mg/kg LD50, skin (Rabbit)

expl. lim.

1.1-7.6 % (v/v)

bp

195-199 °C/1013 hPa

mp

-75 °C

transition temp

flash point 87 °C

density

0.963 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(=O)CCC)C(=O)CCC

InChI

1S/C8H14O3/c1-3-5-7(9)11-8(10)6-4-2/h3-6H2,1-2H3

InChI key

YHASWHZGWUONAO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.966 - 0.968
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service