Skip to Content
Merck
All Photos(1)

Key Documents

65348-M

Sigma-Aldrich

Alcohol 95%

Harleco®, suitable for histology

Synonym(s):

Ethanol, Dehydration, Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116121
application(s):
diagnostic assay manufacturing
histology
bp:
78 °C (lit.)
vapor pressure:
44.6 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

1.59 (vs air)

Quality Level

vapor pressure

44.6 mmHg ( 20 °C)

form

liquid

autoignition temp.

683 °F

shelf life

18 mo. (from manufacture date)

expl. lim.

19 %, 60 °F

manufacturer/tradename

Harleco®

color

APHA: ≤10

refractive index

n20/D 1.3600 (lit.)

bp

78 °C (lit.)

mp

−114 °C (lit.)

application(s)

diagnostic assay manufacturing
histology

storage temp.

10-30°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HARLECO® Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols

Application

HARLECO® Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols
Part of the QSM Histology stain line. Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols.

Analysis Note

Expiration date18 months from mfg dateColor (APHA)10APHA maxFormClear liquidIdentity (IR-spectrum)Conforms to standardSpecific Gravity at 25/25°C0.801-0.817

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service