Skip to Content
Merck
All Photos(1)

Key Documents

573096

Sigma-Aldrich

STAT3 Inhibitor Peptide, Cell-Permeable

≥95% (HPLC), lyophilized solid, STAT3 inhibitor, Calbiochem®

Synonym(s):

STAT3 Inhibitor Peptide, Cell-Permeable, PpYLKTK-mts, STAT3 Inhibitor IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C92H157N20O24P
Molecular Weight:
1958.32
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

STAT3 Inhibitor Peptide, Cell-Permeable, The STAT3 Inhibitor Peptide, Cell-Permeable controls the biological activity of STAT3.

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Supplied as a trifluoroacetate salt.
A cell-permeable analog of the Stat3-SH2 domain-binding phosphopeptide (Cat. No. 573095) that contains a C-terminal mts (membrane translocating sequence) and acts as a highly selective, potent blocker of Stat3 activation. Also suppresses constitutive Stat-3 dependent Src transformation with no effect on Stat-3 independent Ras transformation. The unphosphorylated inactive control peptide is also available under Cat. No. 573105.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Blocker of Stat3 activation
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Pro-Tyr-(PO₃H₂)-Leu-Lys-Thr-Lys-Ala-Ala-Val-Leu-Leu-Pro-Val-Leu-Leu-Ala-Ala-Pro-OH

Other Notes

Turkson, J., et al. 2001. J. Biol. Chem.276, 45443.
Rojas, M., et al. 1998. Nat. Biotechnol.16, 370.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Johansen et al.
The British journal of dermatology, 163(6), 1194-1204 (2010-09-18)
Anti-TNFα therapies are well established for severe psoriasis; however, their mechanism of action in disease resolution is not fully understood. p38 mitogen-activated protein kinase (MAPK) is a kinase known to play a key role in the pathogenesis of psoriasis. To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service