Skip to Content
Merck
All Photos(2)

Key Documents

567741

Sigma-Aldrich

Sphingosine Kinase 1 Inhibitor II, PF-543

The Sphingosine Kinase 1 Inhibitor II, PF-543 controls the biological activity of Sphingosine Kinase 1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Sphingosine Kinase 1 Inhibitor II, PF-543, SK Inhibitor II, ( R)-(1-(4-((3-Methyl-5-(phenylsulfonylmethyl)phenoxy)methyl)benzyl)-pyrrolidin-2-yl)methanol, Sphingosine Kinase Inhibitor II, SK Inhibitor II, (R)-(1-(4-((3-Methyl-5-(phenylsulfonylmethyl)phenoxy)methyl)benzyl)-pyrrolidin-2-yl)methanol, Sphingosine Kinase Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31NO4S
CAS Number:
Molecular Weight:
465.60
Enzyme Commission number:
465.6
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=S(C1=CC=CC=C1)(CC2=CC(C)=CC(OCC3=CC=C(CN4[C@@H](CO)CCC4)C=C3)=C2)=O

InChI

1S/C27H31NO4S/c1-21-14-24(20-33(30,31)27-7-3-2-4-8-27)16-26(15-21)32-19-23-11-9-22(10-12-23)17-28-13-5-6-25(28)18-29/h2-4,7-12,14-16,25,29H,5-6,13,17-20H2,1H3/t25-/m1/s1

InChI key

NPUXORBZRBIOMQ-RUZDIDTESA-N

General description

A cell-permeable hydroxymethylpyrrolidine compound that acts as a high affinity (Kd = 5 nM, kofft1/2 = 8.5 min), but poor, sphingosine kinase-1/SphK1 substrate (1/1,500 of sphingosine processivity) and potently inhibits against SphK1-catalyzed sphingosine-to-S1P conversion (IC50 = 2.0 nM; [sphingosine] = 3 µM) in a reversible, sphingosine-, but not ATP-, competitive manner (Ki = 3.6 nM), exhibiting 132-fold selectivity over SphK2, no affinity toward S1P receptors (S1P1, S1P2, S1P3, S1P5), and much reduced or no inhibitory activity against a panel of 46 other lipid and protein kinases (IC50 >10 µM), including the structurally related diacylglyerol kinase α/DAGKα (3.7% inhibition at 10 µM). PF-543 exposure in 1483 squamous cultures is reported to result in intracellular S1P depletion (by 90% upon 1 h 200 nM or 7 d 1 µM drug exposure) with concomitant elevation of sphingosine level and no detectable cytotoxicity (1 µM for 7 to 9 d in 1483, A549, Jurkat, LN229, MCF7, or U937 cultures). Also active in human whole blood assays in the presence human & bovine serum proteins (IC50 = 26.7 nM against cellular C17-S1P formation from exogenously added 20 µM C17-sphingosine in 10 min with 30 min drug preincubation).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Schnute, M.E., et al. 2012. Biochem. J.444, 79.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service