Skip to Content
Merck
All Photos(1)

Key Documents

566325

Sigma-Aldrich

SIRT1 Inhibitor IV, (S)-35

The SIRT1 Inhibitor IV, (S)-35, also referenced under CAS 848193-72-6, controls the biological activity of SIRT1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

SIRT1 Inhibitor IV, (S)-35, (6 S)-2-Chloro-5,6,7,8,9,10-hexahydro-cyclohepta[b]indole-6-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15ClN2O
CAS Number:
Molecular Weight:
262.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥96% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1cc2c([nH]c3c2CCCC[C@@H]3C(=O)N)cc1

InChI

1S/C14H15ClN2O/c15-8-5-6-12-11(7-8)9-3-1-2-4-10(14(16)18)13(9)17-12/h5-7,10,17H,1-4H2,(H2,16,18)/t10-/m0/s1

InChI key

ABIVOOWWGYJNLV-JTQLQIEISA-N

General description

A cell-permeable entiomerically pure compound that is structurally similar to and exhibits similar potency (IC50 = 63 nM) and selectivity as SIRT1 Inhibitor III (Cat. No. 566322). Shown to be orally bioavailable with a serum half-life of 94 minutes in mice in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Napper, A.D., et al. 2005. J. Med. Chem.48, 8045.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service