Skip to Content
Merck
All Photos(1)

Key Documents

538909

Sigma-Aldrich

Prostaglandin D2

Major prostaglandin metabolite in brain and an important inflammatory mediator in mast cells.

Synonym(s):

Prostaglandin D2, PGD₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 7879

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

ethanol: 100 mg/mL
acetone: soluble
ethyl acetate: soluble
methanol: soluble

shipped in

wet ice

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H]([C@H](C(=O)C1)\C=C\[C@@H](O)CCCCC)C\C=C/CCCC(=O)O

InChI

1S/C20H32O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h4,7,12-13,15-18,21-22H,2-3,5-6,8-11,14H2,1H3,(H,24,25)/b7-4-,13-12+/t15-,16+,17+,18-/m0/s1

InChI key

BHMBVRSPMRCCGG-OUTUXVNYSA-N

General description

Major prostaglandin metabolite in the brain and an important inflammatory mediator in mast cells. Neuromodulator that evokes nerve-mediated Cl- secretion. Stimulates adenylate cyclase.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Adenylate cyclase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).

Other Notes

Horn, E.H., et al. 1995. Clin. Sci. 89, 91.
Frieling, T., et al. 1994. Am. J. Physiol. 266, G132.
Huttemeier, P.C., et al. 1993. Prostaglandins 45, 177.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service