Skip to Content
Merck
All Photos(2)

Key Documents

5.38759

Sigma-Aldrich

MCT1 Inhibitor III

Synonym(s):

MCT1 Inhibitor III, ( E)-2-Cyano-3-(4-(diphenylamino)-2-methoxyphenyl)acrylate, Sodium, Monocarboxylate Transport 1 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H17N2NaO3
Molecular Weight:
392.38
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable, orally bioavailable, water-soluble (~1 mg/ml), non-toxic α-cyanocinnamate analog that acts as a potent and reversible inhibitor of MCT1 activity (IC50 = 11 nM for [14C]-lactate uptake in rat brain endothelial cell line RBE4). Shown to suppress colorectal adenocarcinoma (WiDr cell line) tumor growth in female BALB/c nude mice (% inhibition at the end of 3-weeks = 29, 45, 45 & 56 administered at 10 mg/kg, b.i.d., i.p., 50 mg/kg, b.i.d., p.o., 50 mg/kg, b.i.d., i.p. and 100 mg/kg, q.i.d., p.o., respectively). Displays favorable PK profile in mice (100 mg/kg, p.o.: t1/2 = 3.03 h, Tmax = 0.667 h & Cmax = 17.5 mg/L; 100 mg/kg, i.p.: t1/2 = 3.19 h, Tmax = 0.5 h & Cmax = 61.333 mg/L).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MCT1
Reversible: yes
Target IC50: 11 nM for [¹⁴C]-lactate uptake in rat brain endothelial cell line RBE4

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gurrapu, S., et al. 2015. ACS Med. Chem. Lett.6, 558.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service