Skip to Content
Merck
All Photos(1)

Key Documents

5.08761

Sigma-Aldrich

PCSK9 Inhibitor, EGF-A

Synonym(s):

PCSK9 Inhibitor, EGF-A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C188H292N58O65S6
Molecular Weight:
4597.07
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

potency

0.3 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 25 mg/mL

storage temp.

−20°C

General description

A 42-mer synthetic peptide (GTNECLDNNGGCSHVCNDLKIGYECLCPDGFQLVAQRRCEDI-NH2) that contains a calcium binding site of EGF-A. Binds to the proprotein convertase subtilisin/kexin type 9 (PCSK9) in a pH and calcium-dependent manner (Kd = 300 nM at pH 5.2 and 1.0 µM at pH7.4 and at 2 mM calcium) and blocks its interaction with LDL receptors (IC50 = 3.4 µM) and VLDL receptors (IC50 = 4.7 µM). Acts as a poor inhibitor of PCSK9 - Apo-ER2 interaction even at high concentrations (~200 µM). Blocks the degradation of mature LDL receptors in HepG2 cells in a dose-dependent manner (~1.5 to 15 µM).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
PCSK9

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

GTNECLDNNGGCSHVCNDLKIGYECLCPDGFQLVAQRRCEDI-NH2 (Disulfide Bridge: 5-16, 12-25, 27-39)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Shan L, et al. 2008. Biochem Biophys Res Commun.375, 69.
Da-Wei Zhang, et al. 2007. J. Biol. Chem.282, 18602.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service