Skip to Content
Merck
All Photos(1)

Key Documents

5.08321

Sigma-Aldrich

Dihydroorotate Dehydrogenase Inhibitor, Brequinar

Synonym(s):

Dihydroorotate Dehydrogenase Inhibitor, Brequinar, 6-fluoro-2-(2ʹ-fluoro-1,1ʹ-biphenyl-4-yl)-3-methyl-4-quinoline-carboxylic acid sodium salt, NSC 368390, DuP 785, NSC 368390, DuP 785, 6-fluoro-2-(2ʹ-fluoro-1,1ʹ-biphenyl-4-yl)-3-methyl-4-quinoline-carboxylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H14F2NNaO2
CAS Number:
Molecular Weight:
397.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

12 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[Na+].Fc1c(cccc1)c2ccc(cc2)c3nc4c(c(c3C)C(=O)[O-])cc(cc4)F

InChI

1S/C23H15F2NO2.Na/c1-13-21(23(27)28)18-12-16(24)10-11-20(18)26-22(13)15-8-6-14(7-9-15)17-4-2-3-5-19(17)25;/h2-12H,1H3,(H,27,28);/q;+1/p-1

InChI key

PZOHOALJQOFNTB-UHFFFAOYSA-M

General description

A quinolinecarboxylic acid derivative with immunosuppressive and anti-neoplastic properties. Inhibits pyrimidine synthesis by blocking the activity of mammalian mitochondrial dihydroorotate dehydrogenase (DHODH; IC50 = 12 nM). Does not affect DHODH in non-mammalian species. Also reported to inhibit glycosylation of adhesion molecules. Potentiates the sensitivity of U1690 small cell lung cancer cells to TRAIL-induced apoptosis and synergistically enhances the anti-tumor effects of doxorubicin ( ~100 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DHODH

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

He, T. et al. 2013. Oncogene32, in press.
Hurt, D.E., et al. 2006. Bioorg. Med. Chem. Lett.16, 1610.
Schnellrath, L. et al. 2011. Int. J. Antimicrob. Agents.38, 435.
Batt, D.G., et al. 1995. Bioorg. Med. Chem. Lett.14, 1549.
Allison, A., et al. 1993. Transplantation Proc.25, 67.
Cramer, D. et al. 1992. Transplantation.53, 303.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service