Skip to Content
Merck
All Photos(1)

Key Documents

437720

Sigma-Aldrich

Lipoxin A4

Lipoxin A₄, CAS 89663-86-5, is a potent inhibitor of cytotoxic activity of human natural killer cells. Shown to be as potent as LTB4 in stimulating human neutrophils to generate superoxides.

Synonym(s):

Lipoxin A4, LXA₄, 5(S),6(R),15(S)-Trihydroxyeicosa-7- trans-9- trans-11- cis-13- trans-tetraenoic Acid, LXA₄, 5(S),6(R),15(S)-Trihydroxyeicosa-7-trans-9-trans-11-cis-13-trans-tetraenoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

ethanol: soluble

shipped in

wet ice

storage temp.

−70°C

SMILES string

O[C@H]([C@H](O)\C=C\C=C\C=C/C=C/[C@@H](O)CCCCC)CCCC(=O)O

InChI

1S/C20H32O5/c1-2-3-8-12-17(21)13-9-6-4-5-7-10-14-18(22)19(23)15-11-16-20(24)25/h4-7,9-10,13-14,17-19,21-23H,2-3,8,11-12,15-16H2,1H3,(H,24,25)/b6-4-,7-5+,13-9+,14-10+/t17-,18+,19-/m0/s1

InChI key

IXAQOQZEOGMIQS-SSQFXEBMSA-N

General description

Caution! Product contains volatile liquid. Keep product cold at all times.
Potent inhibitor of cytotoxic activity of human natural killer cells. Shown to be as potent as LTB4 in stimulating human neutrophils to generate superoxides. Involved in contractile responses and acts as a potent enhancer of human protein kinase C. Stimulates the rapid lipid remodeling and pertussis-sensitive release of arachidonic acid in polymorphonuclear leukocytes.
Potent inhibitor of cytotoxic activity of human natural killer cells; shown to be as potent as LTB4 in stimulating human neutrophils to generate superoxides. Involved in contractile responses and acts as a potent activator of human protein kinase C. Stimulates rapid lipid remodeling and pertussis-sensitive release of arachidonic acid in polymorphonuclear leukocytes. Stimulates MAP kinases via activation of G protein-coupled receptors.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Potent inhibitor of cytotoxic activity of human natural killer cells
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Flammable (J)

Physical form

In Ethanol.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
McMahon, B., et al. 2000. J. Biol. Chem. 275, 27566.
Soyombo, O., et al. 1994. Allergy 49, 230.
Flore, S., et al. 1992. J. Biol. Chem.267, 16168.
Badr, K.F., et al. 1989. Proc. Natl. Acad. Sci. USA86, 3438.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Soyombo et al.
Allergy, 49(4), 230-234 (1994-04-01)
Lipoxins are trihydroxytetraene metabolites derived through a double lipoxygenation of arachidonic acid. Lipoxin A4 (LXA4) was prepared by total chemical synthesis, and its capacity to modulate eosinophil migration has been evaluated. LXA4 is a weak and partial chemotactic agent; at
B McMahon et al.
The Journal of biological chemistry, 275(36), 27566-27575 (2000-06-28)
The lipoxygenase-derived eicosanoids leukotrienes and lipoxins are well defined regulators of hemeodynamics and leukocyte recruitment in inflammatory conditions. Here, we describe a novel bioaction of lipoxin A(4) (LXA(4)), namely inhibition of leukotriene D(4) (LTD(4))-induced human renal mesangial cell proliferation, and
K F Badr et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(9), 3438-3442 (1989-05-01)
Lipoxin A4 (LXA4) was competitive with [3H]leukotriene D4 (LTD4) for specific binding to cultured rat glomerular mesangial cells. Half-maximal inhibition was obtained with 100 nM LXA4, compared with 10 nM for unlabeled LTD4. At 10 and 50 nM LXA4 induced
S Fiore et al.
The Journal of biological chemistry, 267(23), 16168-16176 (1992-08-15)
Lipoxin A4 stimulates rapid lipid remodeling and a pertussis toxin-sensitive release of arachidonic acid in polymorphonuclear leukocytes (PMN) (Nigam, S., Fiore, S., Luscinskas, F.W., and Serhan, C.N. (1990) J. Cell. Physiol. 143, 512-523) and has been shown to inhibit leukocyte

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service