Skip to Content
Merck
All Photos(1)

Key Documents

374000

Sigma-Aldrich

Helenalin, A. chamissonis ssp. foliosa

Synonym(s):

Helenalin, A. chamissonis ssp. foliosa

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18O4
CAS Number:
Molecular Weight:
262.30
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white to off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O1[C@H]2[C@H]([C@@H]([C@]3([C@H]([C@@H](C2)C)C=CC3=O)C)O)C(=C)C1=O

InChI

1S/C15H18O4/c1-7-6-10-12(8(2)14(18)19-10)13(17)15(3)9(7)4-5-11(15)16/h4-5,7,9-10,12-13,17H,2,6H2,1,3H3/t7-,9+,10-,12-,13+,15+/m1/s1

InChI key

ZVLOPMNVFLSSAA-XEPQRQSNSA-N

General description

A cell-permeable anti-inflammatory sesquiterpene lactone that acts as a specific NF-κB-DNA binding inhibitor by irreversibly alkylating free sulfhydryls of the cysteine residues on the p65 subunit. Exhibits no effect against the DNA-binding activity of the homodimer of p50 subunit, nor does it affect cellular NF-κB activation/nuclear translocation or IκB dissociation/degradation. An excellent research tool for studying NF-κB-mediated biological processes both in cultures in vitro and in animals in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Boulanger, D., et al. 2007. Vet. Microbiol.119, 330.
Auld, C.A., et al. 2006. Biochem. Biophys. Res. Commun.346, 314.
Huang, P.R., et al. 2005. Cancer Lett.227, 169.
Lyss, G., et al. 1998. J. Biol. Chem.273, 33508.
Lyss, G., et al. 1997. Biol. Chem.378, 951.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service