Skip to Content
Merck
All Photos(1)

Key Documents

346007

Sigma-Aldrich

Glucagon-Like Peptide 1 Receptor Agonist II

Synonym(s):

Glucagon-Like Peptide 1 Receptor Agonist II, 4-(3-(benzyloxy)phenyl)-2-(ethylsulfinyl)-6-(trifluoromethyl)pyrimidine, Glucagon-Like Peptide 1 Receptor Agonist II, Compound B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17F3N2O2S
CAS Number:
Molecular Weight:
406.42
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

>95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

FC(F)(F)c1nc(nc(c1)c2cc(ccc2)OCc3ccccc3)[S](=O)CC

InChI

1S/C20H17F3N2O2S/c1-2-28(26)19-24-17(12-18(25-19)20(21,22)23)15-9-6-10-16(11-15)27-13-14-7-4-3-5-8-14/h3-12H,2,13H2,1H3

InChI key

NTDFYGSSDDMNHI-UHFFFAOYSA-N

General description

A pyrimidine small molecule that acts as a GLP-1 allosteric agonist that induces cAMP signaling in HEK293 cells expressing the GLP-1 receptor with EC50 = 0.66 µM and 99% efficacy (similar data with rat GLP-1 receptor), does not compete with radiolabelled GLP-1 in receptor binding assays, and is not active in cells expressing the GLP-2, GIP, PTH, or glucagon receptors. In rodent islets, insulin secretion increases in a dose-dependent and glucose-dependent manner, either alone or in an additive fashion when combined with the endogenous GLP-1 peptide. In vivo, this molecule exhibits insulinotropic effects when dosed at 10mg/kg in SD rats undergoing an IVGTT and a hyperglycemic clamp. It is also shown to increase insulin secretion in perifusion assays involving normal and diabetic human islets ex vivo, at concentrations between 1 and 10 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sloop, KW., et al. 2010. Diabetes.59, 3099.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service