Skip to Content
Merck
All Photos(1)

Key Documents

341583

Sigma-Aldrich

FGF-2, Basic, Bovine

Synonym(s):

FGF-2, Basic, Bovine, nbbFGF, Fibroblast Growth Factor, Basic, Bovine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

Assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<0.1 ng/μg Endotoxin (ng/μg FGF)

shipped in

ambient

storage temp.

−20°C

General description

Native, bovine basic fibroblast growth factor. A multi-functional factor with chemotactic activity for numerous cell types. A multifunctional factor with chemotactic activity for numerous cell types. Mitogenic to neuroectoderm- and mesoderm-derived cells. Plays a role in the proliferation of neuronal precursors and in neuronal differentiation. A potent angiogenic factor both in vivo and in vitro. Known to activate phopholipase D in endothelial cells. Higher concentrations are found in the brain and pituitary gland. Purified from bovine brain by sequential chromatography.
Native, bovine basic fibroblast growth factor. A multi-functional factor with chemotactic activity for numerous cell types. Mitogenic to neuroectoderm- and mesoderm-derived cells. Plays a role in the proliferation of neuronal precursors and in neuronal differentiation. A potent angiogenic factor both in vivo and in vitro. Activates phospholipase D in endothelial cells.

Biochem/physiol Actions

ED₅₀ = 50-150 pg/ml, in the absence of heparin, for stimulation of ³H-thymidine incorporation by NR6R-3T3 fibroblasts

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered 20 mM Tris-HCl, 1M NaCl, 50 µg BSA/µg FGF, pH 7.0.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C) for long term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -70°C.
Reconstitute to a concentration of ≥10 µg/ml in sterile PBS containing ≥0.1% HSA or BSA.

Other Notes

Ohmiya, M., et al. 2001. J. Neurosci. Res.65, 228.
Murai, N., et al. 1996. J. Neurosurg. 85, 1072.
Thomas, K. 1987. Methods Enzymol.147, 120.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service