Skip to Content
Merck
All Photos(1)

Key Documents

324521

Sigma-Aldrich

Eeyarestatin I

A cell-permeable oxo-imidazolidinyl-hydroxyurea that localizes to ER, where it interacts with AAA ATPase p97 via its nitrofuran-containing moiety, without exhibiting affinity toward Hsp70 / ATPase NSF

Synonym(s):

Eeyarestatin I, 1-(4-Chloro-phenyl)-3-(3-(4-chloro-phenyl)-5,5-dimethyl-1-(3-(5-nitro-furan-2-yl)-allyldiene-hydrazinocarbonylmethyl)-2-oxo-imidazolidin-4-yl)-1-hydroxyl-urea, EerI, ES1, Valosin-containing Protein Inhibitor II, VCP Inhibitor II, ERAD Inhibitor II, p97 Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25Cl2N7O7
CAS Number:
Molecular Weight:
630.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow-orange

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)N2C(C(N(C2=O)CC(=O)NN=CC=Cc4[o]c(cc4)[N+](=O)[O-])(C)C)N(O)C(=O)Nc3ccc(cc3)Cl

InChI

1S/C27H25Cl2N7O7/c1-27(2)24(35(40)25(38)31-19-9-5-17(28)6-10-19)34(20-11-7-18(29)8-12-20)26(39)33(27)16-22(37)32-30-15-3-4-21-13-14-23(43-21)36(41)42/h3-15,24,40H,16H2,1-2H3,(H,31,38)(H,32,37)

InChI key

JTUXTPWYZXWOIB-UHFFFAOYSA-N

General description

A cell-permeable oxo-imidazolidinyl-hydroxyurea that preferentially localizes to ER, where it interacts with AAA (ATPase associated with diverse cellular activities) ATPase p97 (Kd = 5 - 10 µM) via its nitrofuran-containing moiety, without exhibiting affinity toward Hsp70 or AAA ATPase NSF (N-methylmaleimide-sensitive factor). Evidence indicates that EerI cellular metabolite, but not EerI itself, is responsible for the inhibition of ER membrane translocon sec61 complex-mediated transfer of newly synthesized polypeptide, resulting in a blockage of ER-mediated posttranslational glycosylation and signal peptide removal (Effective conc. = 8 µM in HepG2 and HeLa cultures). Ether independent or as a consequence of the early effect on sec61 complex, EerI culture treatment also induces a selective dissociation of an 180 kDa protein from the atx3-containing p97/VCP (Valosin-containing protein) complex and a blockage of the complex-associated deubiquitination of ERAD (ER-associated protein degradation) substrates in a reversible manner, resulting in an accumulation of polyubiquitinated proteins (Effective conc. = 10 µM in A9 and 293T cultures). EerI culture treatment (10 µM) is also demonstrated to selectively induce cytotoxicity in lymphoid cell lines, BJAB, HBL-2, JEKO-1, Jurkat, KMS-12, MINO, as well as primary leukemia cells from CLL (chronic lymphocytic leukemia) patients, but not PBMC from healthy donors, by upregulating the BH3-only pro-apoptotic protein NOXA in cancer cells via ATF3/4 activation and a downregulation of H2A ubiquitination. Unlike DBeQ (Cat. No. 506190), EeRI does not inhibit p97 ATPase activity.
A cell-permeable oxo-imidazolidinyl-hydroxyurea whose cellular metabolite is shown to effectively inhibit the ER membrane translocon sec61 complex-mediated transfer of newly synthesized polypeptide, resulting in a blockage of ER-mediated posttranslational glycosylation and signal peptide removal (Effective conc. = 8 µM in HepG2 and HeLa cultures). Ether independent or as a consequence of the early effect on sec61 complex, EerI culture treatment also induces a selective dissociation of an 180 kDa protein from the atx3-containing p97/VCP (Valosin-containing protein) ATPase complex and a blockage of the p97 complex-associated deubiquitination of ERAD (ER-associated protein degradation) substrates in a reversible manner, resulting in an accumulation of polyubiquitinated proteins (Effective conc. = 10 µM in A9 and 293T cultures). EerI culture treatment (10 µM) is also demonstrated to selectively induce cytotoxicity in lymphoid cell lines, BJAB, HBL-2, JEKO-1, Jurkat, KMS-12, MINO, as well as primary leukemia cells from CLL (chronic lymphocytic leukemia) patients, but not PBMC from healthy donors, by upregulating the BH3-only pro-apoptotic protein NOXA in cancer cells via ATF3/4 activation and a downregulation of H2A ubiquitination.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
It is recommended to prepare a stock solution for 3 month use each time and store the unreconstituted material in solid form, protected from moisture and preferably under inert gas, for best long-term stability during storage.

Other Notes

Chou, T.F., et al. 2011. Proc. Natl. Acad. Sci. USA108, 4834.
Wang. Q, et al. 2010. PLoS ONE5, e15479.
Cross, B.C.S., et al. 2009. J. Cell. Sci.122, 4393.
Wang, Q., et al. 2009. Proc. Natl. Acad. Sci. USA106, 2200.
Wang, Q., et al. 2008. J. Biol. Chem.283, 7445.
Fiebiger, E., et al. 2004. Mol. Biol. Cell15, 1635.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graham Casey et al.
PloS one, 15(11), e0242599-e0242599 (2020-11-24)
Sensorineural hearing loss is one of the most common disabilities worldwide. Such prevalence necessitates effective tools for studying the molecular workings of cochlear cells. One prominent and effective vector for expressing genes of interest in research models is adeno-associated virus
Danielle Bouchard et al.
Molecular biology of the cell, 32(19), 1849-1866 (2021-07-08)
The small ubiquitin-related modifiers (SUMOs) regulate nearly every aspect of cellular function, from gene expression in the nucleus to ion transport at the plasma membrane. In humans, the SUMO pathway has five SUMO paralogues with sequence homologies that range from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service