Skip to Content
Merck
All Photos(1)

Key Documents

2750

Sigma-Aldrich

BrdU Cell Proliferation Kit

The BrdU Cell Proliferation Assay Kit is used to detect the BrdU in a sample.

Synonym(s):

Bromodeoxyuridine Assay, non isotopic assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.76
Pricing and availability is not currently available.

Quality Level

species reactivity

horse, guinea pig, rat, human, canine, mouse, pig, rabbit, hamster, nonhuman primates

packaging

pkg of 200 assay(s)

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

detection method

colorimetric

General description

Test Principle With CHEMICON®′s BrdU Cell Proliferation Assay Kit, mouse monoclonal antibody is used to detect the BrdU in a sample. After addition of goat anti-mouse IgG-peroxidase conjugated secondary antibody, substrate and stop solution, the amount of BrdU is determined. The higher the OD, the higher the BrdU concentration in the sample.

Application

The BrdU Cell Proliferation Assay Kit is used to detect the BrdU in a sample.

The CHEMICON® BrdU Cell Proliferation Assay Kit is a non-isotopic assay for the in vitro quantitative detection of newly synthesized DNA of actively proliferating cells. For Research Use Only. Not for use in diagnostic procedures.

Packaging

200 assays

Components

  • BrdU Reagent: (Part No. 2750a) One 15 μL vial of a 500X solution of BrdU.
  • Fixing Solution: (Part No. 2750b) One 40 mL bottle.
  • Prediluted BrdU Detection Antibody: (Part No. 2750c) One 20 mL vial of an anti-BrdU solution.
  • Stop Solution: (Part No. 2750d): One 25 mL bottle of 2.5N sulfuric acid solution.
  • Goat anti-Mouse IgG, Peroxidase labeled: (Part No. 2750e) One 15 μL vial of a 2000X solution.
  • Conjugate Diluent: (Part No. 2750f) One 25 mL bottle.
  • Substrate: (Part No. 2750g) One 25 mL bottle, ready to use TMB.
  • 50X Plate Wash Concentrate: (Part No. 2750h) One 90 mL bottle of buffered Tris with surfactant.

Storage and Stability

Maintain the kit at 2° to 8°C under sterile conditions for up to 6 months after date of receipt.

Before first use: remove the Fixative Solution and place at room temperature for at least 4 hours prior to use. Precipitates that may occur during cold storage should go back into solution.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service