Skip to Content
Merck
All Photos(1)

Key Documents

265005

Sigma-Aldrich

Dexamethasone

≥99% (TLC), solid, NOS induction inhibitor, Calbiochem

Synonym(s):

Dexamethasone, 9α-Fluoro-16α-methylprednisolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
MDL number:
UNSPSC Code:
51422306
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Dexamethasone, Most active and highly stable glucocorticoid.

description

Merck USA index - 14, 2943

Quality Level

Assay

≥99% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 25 mg/mL
chloroform: soluble
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

F[C@@]21[C@H]([C@H]4[C@@]([C@@]([C@@H](C4)C)(O)C(=O)CO)(C[C@@H]2O)C)CCC3=CC(=O)C=C[C@@]31C

InChI

1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1

InChI key

UREBDLICKHMUKA-CXSFZGCWSA-N

General description

Most active and highly stable glucocorticoid. Causes a reduction in cyclin A and Cdk2 activity, and inhibition of G1/S transition in osteoblasts. Inhibits the phosphorylation of retinoblastoma (Rb) protein. Inhibits the expression of inducible but not constitutive nitric oxide synthase in vascular endothelial cells (IC50 = 5 nM). Enhances active cation transport in aortic smooth muscle cells by stimulating the Na+ - K+ pump. Induces apoptosis in human thymocytes.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Phosphorylation of rb protein
Product does not compete with ATP.
Reversible: no
Target IC50: 5 nM Inhibiting the expression of inducible but not constitutive nitric oxide synthase in vascular endothelial cells

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Reil, T.D., et al. 2000. J. Surg. Res.92, 108.
Smith, E., et al. 2000. J.Biol. Chem. 275, 1992.
Petit, P.X., et al. 1995. J. Cell Biol.130, 157.
Stern, N., et al. 1994. Am. J. Hypertens. 7, 146.
Donis, J.A., et al. 1993. Biotechniques15, 786.
Ohoka, Y., et al. 1993. Biochem. Biophys. Res. Commun.197, 916.
Sarin, A., et al. 1993. J. Exp. Med.178, 1693.
Vandewalle, A., et al. 1993. J. Cell Physiol.156, 383.
Nieto, M.A., et al. 1992. Clin. Exp. Immunol.88, 341.
Walsh, D., and Avashia, J. 1992. Cleveland Clin. J. Med. 59, 505.
Warters, R.L. 1992. Cancer Res.52, 883.
Sperber, K., et al. 1991. J. Allergy Clin. Immunol.87, 490.
Radomski, M.W., et al. 1990. Proc. Natl. Acad. Sci. USA 87, 10043.
Bettger, W.J., et al. 1981. Proc. Natl. Acad. Sci. USA78, 5588.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arif Yurdagul et al.
Cell metabolism, 31(3), 518-533 (2020-02-01)
Continual efferocytic clearance of apoptotic cells (ACs) by macrophages prevents necrosis and promotes injury resolution. How continual efferocytosis is promoted is not clear. Here, we show that the process is optimized by linking the metabolism of engulfed cargo from initial

Questions

  1. What is the solubility ratio of your product with the code 265005 dexamethasone in chloroform and ethanol, how many milligrams in how many milliliters. Additionally, does it need a sonicator or heat temperature to dissolve?

    1 answer
    1. Dexamethasone can be solubilized in ethanol or methanol at 25 mg/mL. This chemical has been cited by various sources as slightly soluble in chloroform, however the rate has not been reported. Alternatively, product D2915 is a Dexamethasone product containing cyclodextrin. This product is water soluble. See the link below for additional information:
      https://www.sigmaaldrich.com/product/sigma/d2915

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service