Skip to Content
Merck
All Photos(1)

Key Documents

219392

Sigma-Aldrich

Cathepsin B Substrate III, Fluorogenic

≥95% (HPLC), lyophilized solid, Cathepsin B inhibitor, Calbiochem®

Synonym(s):

Cathepsin B Substrate III, Fluorogenic, Z-RR-AMC, 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H39N9O6 · 2HCl
Molecular Weight:
694.61
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Cathepsin B Substrate III, Fluorogenic, Sensitive fluorogenic substrate for the quantitative determination of cathepsin B activity.

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 5 mg/mL
water: soluble

fluorescence

λex 360-380 nm
λem 430-460 nm

shipped in

ambient

storage temp.

−20°C

General description

Sensitive fluorogenic substrate for the quantitative determination of cathepsin B activity (excitation maximum: 380 nm; emission maximum: 460 nm).

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Arg-Arg-AMC, 2HCl

Other Notes

Hiwasa, T., et al. 1987. FEBS Lett.211, 23.
Manson, R.W., et al. 1985. Biochem. J.226, 233.
Tanaka, M., et al. 1985. Life Sci. 36, 1217.
Docherty, K., et al. 1984. J. Biol. Chem.259, 6041.
Barrett, A.J., and Kirschke, H. 1981. Methods Enzymol.80, 535.
Barrett, A.J. 1980. Biochem. J.187, 909.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Cui et al.
Frontiers in cell and developmental biology, 8, 31-31 (2020-03-03)
Lysosomal membrane permeabilization (LMP) has recently been recognized as an important cell death pathway in various cell types. However, studies regarding the correlation between LMP and cardiomyocyte death are scarce. Lysosomal membrane-associated protein 2 (Lamp2) is an important component of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service