Skip to Content
Merck
All Photos(1)

Key Documents

212385-M

Millipore

Carbachol

An unsubstituted carbamoyl ester that acts as an agonist for nicotinic and muscarinic acetylcholine receptors.

Synonym(s):

Carbachol, Carbamoylcholine Chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15ClN2O2
CAS Number:
Molecular Weight:
182.65
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 1779

Assay

≥98% (titration)

form

solid (fine crystalline)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

methanol: 100 mg/mL
water: 100 mg/mL
ethanol: 20 mg/mL

storage temp.

10-30°C

SMILES string

Cl.[N+](CCO\C(=N\[H])\[O-])(C)(C)C

InChI

1S/C6H14N2O2.ClH/c1-8(2,3)4-5-10-6(7)9;/h4-5H2,1-3H3,(H-,7,9);1H

InChI key

AIXAANGOTKPUOY-UHFFFAOYSA-N

General description

An unsubstituted carbamoyl ester that acts as an agonist for nicotinic and muscarinic acetylcholine receptors. Known to increase cytosolic Ca2+ and InsP3 and decrease cAMP levels. Resistant to the action of acetylcholine esterase (AChE) and non-specific cholinesterases. Inhibits apoptotic death of cultured granule neurons.

Biochem/physiol Actions

Primary Target
Agonist for nicotinic and muscarinic acetylcholine receptors

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Jence, M., et al. 1995. J. Neurochem. 64, 1605.
Yan, G.M., et al. 1995. Mol. Pharmacol. 47, 248.
Mangels, L.A., and Gnegy, M.E. 1992. J. Biol. Chem.267, 5847.
Watanabe, A.M., et al. 1992. In Basic and Clinical Pharmacology, Fifth Edition Katzung, B.G. ed.) Appleton and Lange, Norwalk, CT, p. 82.
Howe, P.H., et al. 1986. J. Pharmacol. Exp. Ther.239, 574.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service