Skip to Content
Merck
All Photos(1)

Key Documents

1.93418

Supelco

Oxalic acid dihydrate

EMPLURA®

Synonym(s):

Oxalic acid dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COOH)2 · 2 H2O
CAS Number:
Molecular Weight:
126.07
MDL number:
UNSPSC Code:
12352106

product line

EMPLURA®

storage temp.

room temp

InChI

1S/C2H2O4.2H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H2

InChI key

GEVPUGOOGXGPIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

1.93418.0521: 500 g, Minimum order quantity: 20
1.93418.5021: 5 kg, Minimum order quantity: 4
1.93418.9051: 50 kg, Minimum order quantity: 1

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Inorganics and solvents for basic and standard applications manufactured locally.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service