Skip to Content
Merck
All Photos(1)

Key Documents

182516-M

Millipore

Arp2/3 Complex Inhibitor II, CK-869

The Arp2/3 Complex Inhibitor II, CK-869 controls the biological activity of Arp2/3. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Arp2/3 Complex Inhibitor II, CK-869, Actin Assembly Inhibitor XIV, 2-(3-Bromophenyl)-3-(2,4-dimethoxyphenyl)-1,3-thiazolidin-4-one, racemic, CK-0157869

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16BrNO3S
Molecular Weight:
394.28
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable thiazolidinone compound that inhibits actin assembly mediated by actin-related protein Arp2/3 complex of human and bovine, but not yeast, species (IC50 = 11 µM against bovine Arp2/3). Shown to inhibit the actin filament "comet tails" formation around intracellular Listeria in infected SKOV3 cells (IC50 = 7 µM). Structrual studies indicate that CK-869 targets the hydrophobic core in subdomain 1 of Arp3. CK-666 (Cat. No. 182515) and CK-869 exhibit different modes of binding, resulting in their different yeast cross-reactivities. Supplied as a racemic mixture of the active 2S and the inactive 2R enantiomers. CK-312 (Cat. No. 182518) can serve as a negative control.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Nolen, B.J., et al. 2009. Nature460, 1031.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service